siRNA Details
| Gene Symbol: Slc6a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172321.1 | NP_758824.1 | 3 | 1213 | ||
| GenBank mRNA | L22022.1 | - | 3 | 1213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a15| Gene Symbol: Slc6a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172321.1 | NP_758824.1 | 3 | 1213 | ||
| GenBank mRNA | L22022.1 | - | 3 | 1213 | ||