siRNA Details
| Gene Symbol: Pycard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172322.1 | NP_758825.1 | 1 | 260 | ||
| GenBank mRNA | AB053165.1 | - | 1 | 260 | ||
| FQ226280.1 | - | 1 | 418 | |||
| FQ227544.1 | - | 1 | 439 | |||
| FQ227755.1 | - | 1 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pycard| Gene Symbol: Pycard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172322.1 | NP_758825.1 | 1 | 260 | ||
| GenBank mRNA | AB053165.1 | - | 1 | 260 | ||
| FQ226280.1 | - | 1 | 418 | |||
| FQ227544.1 | - | 1 | 439 | |||
| FQ227755.1 | - | 1 | 418 | |||