siRNA Details
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172323.1 | NP_758826.1 | 5 | 1720 | ||
| GenBank mRNA | AB097568.1 | - | 5 | 1720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Has1| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172323.1 | NP_758826.1 | 5 | 1720 | ||
| GenBank mRNA | AB097568.1 | - | 5 | 1720 | ||