siRNA Details
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172326.1 | NP_758829.1 | 22 | 2153 | ||
GenBank mRNA | AY149902.1 | - | 22 | 2153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172326.1 | NP_758829.1 | 22 | 2153 | ||
GenBank mRNA | AY149902.1 | - | 22 | 2153 |