siRNA Details
Gene Symbol: Zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172330.3 | NP_758833.2 | 9, 10 | 1356 | ||
GenBank mRNA | AF456325.1 | - | 9, 10 | 1357 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172330.3 | NP_758833.2 | 9, 10 | 1356 | ||
GenBank mRNA | AF456325.1 | - | 9, 10 | 1357 |