siRNA Details
| Gene Symbol: Zp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172330.3 | NP_758833.2 | 5 | 728 | ||
| GenBank mRNA | AF456325.1 | - | 5 | 729 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zp4| Gene Symbol: Zp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172330.3 | NP_758833.2 | 5 | 728 | ||
| GenBank mRNA | AF456325.1 | - | 5 | 729 | ||