siRNA Details
| Gene Symbol: Fibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172334.2 | NP_758837.1 | 6 | 738 | ||
| XM_006230701.2 | XP_006230763.1 | 6 | 815 | |||
| GenBank mRNA | AY093421.1 | - | 6 | 739 | ||
| BC061802.1 | - | 6 | 759 | 5598566 | ||
| FQ228181.1 | - | 6 | 790 | |||
| FQ234469.1 | - | 6 | 790 | |||
| FQ234564.1 | - | 6 | 790 | |||
| FQ235188.1 | - | 5 | 715 | |||

Fibp
Genomic Map