siRNA Details
| Gene Symbol: Mc2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100491.1 | NP_001093961.1 | 2 | 676 | ||
| XM_006254893.2 | XP_006254955.1 | 2 | 724 | |||
| XM_008772091.1 | XP_008770313.1 | 3 | 775 | |||
| XM_008772092.1 | XP_008770314.1 | 3 | 771 | |||
| XM_008772093.1 | XP_008770315.1 | 3 | 1769 | |||
| XM_008772094.1 | XP_008770316.1 | 2 | 1718 | |||
| XM_008772095.1 | XP_008770317.1 | 3 | 1722 | |||
| GenBank mRNA | AF547168.1 | - | 1 | 334 | ||
More Information
Set Membership: |
|

Mc2r
Genomic Map