siRNA Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005762.1 | NP_001005762.1 | 9 | 948 | ||
| XM_006252241.2 | XP_006252303.1 | 10 | 2632 | |||
| XM_006252242.1 | XP_006252304.1 | 9 | 1039 | |||
| XM_006252244.2 | XP_006252306.1 | 8 | 1267 | |||
| XM_006252246.1 | XP_006252308.1 | 9 | 984 | |||
| XM_008770819.1 | XP_008769041.1 | 7 | 2741 | |||
| GenBank mRNA | AF439378.1 | - | 8 | 764 | ||
| BC083666.1 | - | 9 | 948 | 7135547 | ||

Sorbs3
Genomic Map