siRNA Details
| Gene Symbol: Aqp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173105.1 | NP_775128.1 | 2 | 924 | ||
| GenBank mRNA | AB023644.1 | - | 2 | 924 | ||
| BC078694.1 | - | 2 | 951 | 7111950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp11| Gene Symbol: Aqp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173105.1 | NP_775128.1 | 2 | 924 | ||
| GenBank mRNA | AB023644.1 | - | 2 | 924 | ||
| BC078694.1 | - | 2 | 951 | 7111950 | ||