siRNA Details
| Gene Symbol: Csn1s2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173106.1 | NP_775129.1 | 7, 8 | 445 | ||
| GenBank mRNA | AY154894.1 | - | 7, 8 | 445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csn1s2b| Gene Symbol: Csn1s2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173106.1 | NP_775129.1 | 7, 8 | 445 | ||
| GenBank mRNA | AY154894.1 | - | 7, 8 | 445 | ||