siRNA Details
| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176857.3 | NP_789827.3 | 1 | 1330 | ||
| GenBank mRNA | AB045378.2 | - | 1 | 1330 | ||
| AF289091.1 | - | 1388 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcpip1| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176857.3 | NP_789827.3 | 1 | 1330 | ||
| GenBank mRNA | AB045378.2 | - | 1 | 1330 | ||
| AF289091.1 | - | 1388 | ||||