siRNA Details
Gene Symbol: Vom1r101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173112.2 | NP_775135.2 | 1 | 584 | ||
GenBank mRNA | AY510287.1 | - | 1 | 584 | ||
U36786.1 | - | 1 | 502 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173112.2 | NP_775135.2 | 1 | 584 | ||
GenBank mRNA | AY510287.1 | - | 1 | 584 | ||
U36786.1 | - | 1 | 502 |