siRNA Details
| Gene Symbol: Vom1r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173113.1 | NP_775136.1 | 2 | 1346 | ||
| GenBank mRNA | U36785.1 | - | 2 | 1346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r105| Gene Symbol: Vom1r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173113.1 | NP_775136.1 | 2 | 1346 | ||
| GenBank mRNA | U36785.1 | - | 2 | 1346 | ||