siRNA Details
| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173121.1 | NP_775144.1 | 2 | 506 | ||
| XM_006249965.2 | XP_006250027.1 | 2 | 924 | |||
| GenBank mRNA | AB077854.1 | - | 2 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brinp3| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173121.1 | NP_775144.1 | 2 | 506 | ||
| XM_006249965.2 | XP_006250027.1 | 2 | 924 | |||
| GenBank mRNA | AB077854.1 | - | 2 | 506 | ||