siRNA Details
Gene Symbol: Brinp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173121.1 | NP_775144.1 | 3 | 796 | ||
XM_006249965.2 | XP_006250027.1 | 3 | 1214 | |||
GenBank mRNA | AB077854.1 | - | 3 | 796 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Brinp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173121.1 | NP_775144.1 | 3 | 796 | ||
XM_006249965.2 | XP_006250027.1 | 3 | 1214 | |||
GenBank mRNA | AB077854.1 | - | 3 | 796 |