siRNA Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004442.1 | NP_001004442.1 | 8, 9 | 870 | ||
| GenBank mRNA | BC082063.1 | - | 8, 9 | 870 | 7132589 | |
| FQ229584.1 | - | 3, 4 | 328 | |||
| X79328.1 | - | 7, 8 | 810 | |||

Pdia6
Genomic Map