siRNA Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173125.1 | NP_775148.1 | 9, 10 | 940 | ||
| XM_006253610.2 | XP_006253672.2 | 9, 10 | 1131 | |||
| XM_008771487.1 | XP_008769709.1 | 9, 10 | 1233 | |||
| GenBank mRNA | AF095585.1 | - | 9, 10 | 940 | ||
| BC078693.1 | - | 9, 10 | 932 | 7121395 | ||

Pdlim7
Genomic Map