siRNA Details
| Gene Symbol: Smim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173126.3 | NP_775149.1 | 2 | 1480 | ||
| XM_006254762.2 | XP_006254824.1 | 4 | 2163 | |||
| XM_006254763.2 | XP_006254825.1 | 4 | 2166 | |||
| XM_006254765.2 | XP_006254827.1 | 4 | 1692 | |||
| XM_006254767.1 | XP_006254829.1 | 2 | 1166 | |||
| XM_006254769.2 | XP_006254831.1 | 2 | 1369 | |||
| XM_008772096.1 | XP_008770318.1 | 3 | 2294 | |||
| XM_008772097.1 | XP_008770319.1 | 2 | 2102 | |||
| GenBank mRNA | AF313411.1 | - | 2 | 1278 | ||
| BC070515.1 | - | 2 | 1479 | 7100997 | ||

Smim3
Genomic Map