siRNA Details
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173127.1 | NP_775150.1 | 4 | 517 | ||
GenBank mRNA | FQ230379.1 | - | 4 | 548 | ||
FQ235277.1 | - | 4 | 548 | |||
M16624.1 | - | 4 | 517 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173127.1 | NP_775150.1 | 4 | 517 | ||
GenBank mRNA | FQ230379.1 | - | 4 | 548 | ||
FQ235277.1 | - | 4 | 548 | |||
M16624.1 | - | 4 | 517 |