siRNA Details
| Gene Symbol: Cdk5rap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173134.2 | NP_775157.2 | 15 | 1787 | ||
| XM_006238273.2 | XP_006238335.1 | 15 | 2017 | |||
| XM_006238274.2 | XP_006238336.1 | 15 | 2017 | |||
| XM_006238276.2 | XP_006238338.1 | 15 | 2015 | |||
| XM_006238277.2 | XP_006238339.1 | 15 | 2015 | |||
| XM_006238278.2 | XP_006238340.1 | 15 | 2015 | |||
| XM_006238279.2 | XP_006238341.1 | 15 | 2014 | |||
| XM_006238280.2 | XP_006238342.1 | 15 | 2015 | |||
| XM_008763760.1 | XP_008761982.1 | 15 | 2017 | |||
| XM_008763761.1 | XP_008761983.1 | 15 | 2018 | |||
| GenBank mRNA | JX524852.1 | - | 15 | 1631 | ||

Cdk5rap2
Genomic Map