siRNA Details
| Gene Symbol: Cryba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173140.1 | NP_775163.1 | 4 | 400 | ||
| GenBank mRNA | AY158891.1 | - | 4 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryba2| Gene Symbol: Cryba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173140.1 | NP_775163.1 | 4 | 400 | ||
| GenBank mRNA | AY158891.1 | - | 4 | 400 | ||