siRNA Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173142.1 | NP_775165.1 | 5 | 787 | ||
| XM_006245160.1 | XP_006245222.1 | 5 | 790 | |||
| GenBank mRNA | D86220.1 | - | 5 | 787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk16| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173142.1 | NP_775165.1 | 5 | 787 | ||
| XM_006245160.1 | XP_006245222.1 | 5 | 790 | |||
| GenBank mRNA | D86220.1 | - | 5 | 787 | ||