siRNA Details
| Gene Symbol: Stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173142.1 | NP_775165.1 | 3 | 431 | ||
| XM_006245160.1 | XP_006245222.1 | 3 | 434 | |||
| GenBank mRNA | D86220.1 | - | 3 | 431 | ||
| FQ213827.1 | - | 3 | 427 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AB022179.

Stk16
Genomic Map