siRNA Details
| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173146.2 | NP_775169.4 | 3109 | |||
| XM_008766434.1 | XP_008764656.1 | 3 | 1002 | |||
| GenBank mRNA | U75361.1 | - | 3123 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13c| Gene Symbol: Unc13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173146.2 | NP_775169.4 | 3109 | |||
| XM_008766434.1 | XP_008764656.1 | 3 | 1002 | |||
| GenBank mRNA | U75361.1 | - | 3123 | |||