siRNA Details
| Gene Symbol: SEC62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003262.3 | NP_003253.1 | 3 | 263 | ||
| XM_011513114.2 | XP_011511416.1 | 4 | 340 | |||
| GenBank mRNA | BC003677.1 | - | 3 | 255 | ||
| BC007040.1 | - | 3 | 255 | |||
| BC012035.1 | - | 3 | 247 | 4706586 | ||
| BC105971.1 | - | 346 | ||||
| BX475495.1 | - | 3 | 264 | |||
| BX648539.1 | - | 3 | 247 | |||
| D87127.1 | - | 3 | 244 | |||
| KC005990.1 | - | 3 | 232 | |||
| U93239.1 | - | 3 | 244 | |||

SEC62
Genomic Map