siRNA Details
| Gene Symbol: Vps54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173147.1 | NP_775170.1 | 8 | 1202 | ||
| GenBank mRNA | AJ010392.3 | - | 8 | 1202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps54| Gene Symbol: Vps54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173147.1 | NP_775170.1 | 8 | 1202 | ||
| GenBank mRNA | AJ010392.3 | - | 8 | 1202 | ||