siRNA Details
| Gene Symbol: Obp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173148.1 | NP_775171.1 | 376 | |||
| XM_008761579.1 | XP_008759801.1 | 5 | 481 | |||
| GenBank mRNA | M76734.1 | - | 376 | |||
| Gene Symbol: LOC103691751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761619.1 | XP_008759841.1 | 382 | |||

Obp2b
Genomic Map