siRNA Details
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173153.2 | NP_775176.2 | 3 | 491 | ||
| GenBank mRNA | AM285343.1 | - | 3 | 491 | ||
| AM285683.1 | - | 3 | 491 | |||
| AY070268.1 | - | 2 | 473 | |||
| BC070952.1 | - | 3 | 479 | 7100695 | ||
| CK479589.1 | - | 3 | 491 | |||
| DQ125339.1 | - | 3 | 480 | |||
| DQ125340.1 | - | 3 | 480 | |||

Gimap4
Genomic Map