siRNA Details
| Gene Symbol: Vom1r102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173298.2 | NP_775420.2 | 1 | 893 | ||
| GenBank mRNA | AY510305.1 | - | 1 | 893 | ||
| U36899.1 | - | 3 | 1362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r102| Gene Symbol: Vom1r102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173298.2 | NP_775420.2 | 1 | 893 | ||
| GenBank mRNA | AY510305.1 | - | 1 | 893 | ||
| U36899.1 | - | 3 | 1362 | |||