siRNA Details
| Gene Symbol: LOC286960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173301.1 | NP_775423.1 | 2, 3 | 209 | ||
| GenBank mRNA | X15679.1 | - | 2, 3 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC286960| Gene Symbol: LOC286960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173301.1 | NP_775423.1 | 2, 3 | 209 | ||
| GenBank mRNA | X15679.1 | - | 2, 3 | 209 | ||