siRNA Details
| Gene Symbol: Abca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173307.1 | NP_775429.1 | 33, 34 | 4273 | ||
| XM_006247560.2 | XP_006247622.1 | 32, 33 | 4507 | |||
| GenBank mRNA | AJ426052.2 | - | 33, 34 | 4273 | ||
| AJ550165.1 | - | 32, 33 | 4262 | |||

Abca5
Genomic Map