siRNA Details
| Gene Symbol: Fut11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173308.2 | NP_775430.2 | 3 | 1456 | ||
| GenBank mRNA | AJ535753.1 | - | 3 | 1481 | ||
| BC079316.1 | - | 3 | 1456 | 7135022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut11| Gene Symbol: Fut11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173308.2 | NP_775430.2 | 3 | 1456 | ||
| GenBank mRNA | AJ535753.1 | - | 3 | 1481 | ||
| BC079316.1 | - | 3 | 1456 | 7135022 | ||