siRNA Details
| Gene Symbol: Elavl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238385.2 | XP_006238447.1 | 6 | 786 | ||
| XM_006238388.2 | XP_006238450.1 | 5 | 589 | |||
| XM_006238389.2 | XP_006238451.1 | 5 | 1126 | |||
| XM_006238391.2 | 5 | 1126 | ||||
| XM_006238395.2 | XP_006238457.1 | 4 | 517 | |||
| XM_008763848.1 | XP_008762070.1 | 4 | 832 | |||
| XM_008763849.1 | XP_008762071.1 | 4 | 832 | |||
| XM_008763850.1 | XP_008762072.1 | 4 | 495 | |||
| GenBank mRNA | AB098713.1 | - | 5 | 1050 | ||

Elavl2
Genomic Map