siRNA Details
| Gene Symbol: Chst15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173310.3 | NP_775432.3 | 5 | 1235 | ||
| XM_006230430.2 | XP_006230492.1 | 6 | 2802 | |||
| XM_006230431.2 | XP_006230493.1 | 6 | 3542 | |||
| XM_006230433.2 | XP_006230495.1 | 6 | 3037 | |||
| XM_008759868.1 | XP_008758090.1 | 6 | 1798 | |||
| GenBank mRNA | AB086953.1 | - | 5 | 1235 | ||

Chst15
Genomic Map