siRNA Details
| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173314.2 | NP_775436.2 | 4 | 1199 | ||
| XM_006232101.2 | XP_006232163.1 | 4 | 1435 | |||
| GenBank mRNA | BC161833.1 | - | 5 | 1031 | ||
| U67083.1 | - | 1187 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp455| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173314.2 | NP_775436.2 | 4 | 1199 | ||
| XM_006232101.2 | XP_006232163.1 | 4 | 1435 | |||
| GenBank mRNA | BC161833.1 | - | 5 | 1031 | ||
| U67083.1 | - | 1187 | ||||