siRNA Details
Gene Symbol: LOC286987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173321.1 | NP_775443.1 | 538 | |||
GenBank mRNA | M60388.1 | - | 538 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC286987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173321.1 | NP_775443.1 | 538 | |||
GenBank mRNA | M60388.1 | - | 538 |