siRNA Details
| Gene Symbol: Pnrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173322.1 | NP_775444.1 | 2 | 780 | ||
| GenBank mRNA | FQ214116.1 | - | 2 | 822 | ||
| U61729.1 | - | 2 | 780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnrc1| Gene Symbol: Pnrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173322.1 | NP_775444.1 | 2 | 780 | ||
| GenBank mRNA | FQ214116.1 | - | 2 | 822 | ||
| U61729.1 | - | 2 | 780 | |||