siRNA Details
| Gene Symbol: Tox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173324.2 | NP_775446.2 | 7 | 1278 | ||
| GenBank mRNA | AF267197.1 | - | 6 | 1230 | ||
| BC081696.1 | - | 7 | 1278 | 7112549 | ||
| FQ210274.1 | - | 7 | 1286 | |||
| FQ212245.1 | - | 7 | 1286 | |||

Tox4
Genomic Map