siRNA Details
| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173327.1 | NP_775449.1 | 18 | 2447 | ||
| GenBank mRNA | AF084205.1 | - | 18 | 2447 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taok1| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173327.1 | NP_775449.1 | 18 | 2447 | ||
| GenBank mRNA | AF084205.1 | - | 18 | 2447 | ||