siRNA Details
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173328.1 | NP_775450.1 | 11 | 1046 | ||
| GenBank mRNA | AF061443.1 | - | 11 | 1046 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgr4| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173328.1 | NP_775450.1 | 11 | 1046 | ||
| GenBank mRNA | AF061443.1 | - | 11 | 1046 | ||