siRNA Details
| Gene Symbol: Defa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173329.2 | NP_775451.2 | 3 | 425 | ||
| XM_006253367.1 | XP_006253429.1 | 3 | 469 | |||
| GenBank mRNA | U16685.1 | - | 3 | 404 | ||
| U16686.1 | - | 3 | 425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defa5| Gene Symbol: Defa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173329.2 | NP_775451.2 | 3 | 425 | ||
| XM_006253367.1 | XP_006253429.1 | 3 | 469 | |||
| GenBank mRNA | U16685.1 | - | 3 | 404 | ||
| U16686.1 | - | 3 | 425 | |||