siRNA Details
| Gene Symbol: Mapk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173331.2 | NP_775453.1 | 9 | 906 | ||
| XM_006241777.2 | XP_006241839.1 | 10 | 1322 | |||
| XM_006241779.2 | XP_006241841.1 | 10 | 1322 | |||
| XM_008765547.1 | XP_008763769.1 | 10 | 1174 | |||
| GenBank mRNA | AF078798.2 | - | 9 | 1022 | ||
| FQ212754.1 | - | 4 | 786 | |||

Mapk15
Genomic Map