siRNA Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173332.2 | NP_775454.2 | 7 | 1346 | ||
| XM_006237141.2 | XP_006237203.1 | 8 | 1869 | |||
| XM_006237142.2 | XP_006237204.1 | 7 | 2925 | |||
| XM_006237143.2 | XP_006237205.1 | 7 | 1723 | |||
| XM_006237145.2 | XP_006237207.1 | 8 | 2186 | |||
| XM_006237146.1 | XP_006237208.1 | 5 | 1027 | |||
| XM_008763224.1 | XP_008761446.1 | 8 | 2187 | |||
| XM_008763225.1 | XP_008761447.1 | 9 | 1940 | |||
| XM_008763226.1 | XP_008761448.1 | 7 | 2040 | |||
| XM_008763227.1 | XP_008761449.1 | 5 | 1154 | |||
| GenBank mRNA | BC162051.1 | - | 7 | 1346 | 7112072 | |

Zfand4
Genomic Map