siRNA Details
| Gene Symbol: Olr1361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173333.1 | NP_775455.1 | 1 | 884 | ||
| GenBank mRNA | M64377.1 | - | 1 | 884 | ||
| Gene Symbol: Olr1358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214822.1 | NP_999987.1 | 1 | 884 | ||

Olr1361
Genomic Map