siRNA Details
| Gene Symbol: Olr287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173334.1 | NP_775456.1 | 1 | 315 | ||
| GenBank mRNA | M64378.1 | - | 1 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr287| Gene Symbol: Olr287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173334.1 | NP_775456.1 | 1 | 315 | ||
| GenBank mRNA | M64378.1 | - | 1 | 315 | ||