siRNA Details
| Gene Symbol: Nubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009619.1 | NP_001009619.1 | 7 | 495 | ||
| GenBank mRNA | BC088221.1 | - | 7 | 495 | 7321552 | |
| FQ214074.1 | - | 6 | 439 | |||
| FQ218444.1 | - | 7 | 508 | |||
| FQ220638.1 | - | 7 | 508 | |||
| FQ222868.1 | - | 7 | 512 | |||
| FQ229330.1 | - | 7 | 511 | |||

Nubp1
Genomic Map