siRNA Details
| Gene Symbol: Clec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767505.1 | XP_008765727.1 | 16 | 1991 | ||
| XM_008767506.1 | XP_008765728.1 | 15 | 1985 | |||
| XM_008767507.1 | XP_008765729.1 | 16 | 1943 | |||
| XM_008767508.1 | XP_008765730.1 | 15 | 1937 | |||
| XM_008767509.1 | XP_008765731.1 | 15 | 1937 | |||

Clec16a
Genomic Map