siRNA Details
| Gene Symbol: TSPAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004615.3 | NP_004606.2 | 2 | 274 | ||
| GenBank mRNA | AB062057.1 | - | 2 | 275 | ||
| AB102665.1 | - | 2 | 190 | |||
| AK293178.1 | - | 3 | 259 | |||
| AK293534.1 | - | 4 | 428 | |||
| AK293552.1 | - | 3 | 339 | |||
| AK312343.1 | - | 2 | 208 | |||
| BC018036.1 | - | 2 | 233 | 4801388 | ||
| D10653.1 | - | 2 | 214 | |||
| D29808.1 | - | 2 | 200 | |||
| L10373.1 | - | 2 | 258 | |||

TSPAN7
Genomic Map