siRNA Details
| Gene Symbol: Rogdi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024864.1 | NP_001020035.1 | 10, 11 | 1008 | ||
| GenBank mRNA | BC083789.1 | - | 8, 9 | 1439 | ||
| BC098001.1 | - | 10, 11 | 1008 | 7453517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rogdi| Gene Symbol: Rogdi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024864.1 | NP_001020035.1 | 10, 11 | 1008 | ||
| GenBank mRNA | BC083789.1 | - | 8, 9 | 1439 | ||
| BC098001.1 | - | 10, 11 | 1008 | 7453517 | ||